missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD46 Protein

A DNA sequence encoding the Mus musculus (Mouse) CD46, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5790-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987769

Quantity Price
1 639.01 € / 100µg
5 601.25 € / 100µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_034908.1
CD46 Protein
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
17221
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cd46
Recombinant Protein
E. coli
CELPRPFEAM ELKGTPKLFY AVGEKIEYKC KKGYLYLSPY LMIATCEPNH TWVPISDAGC IKVQCTMLQD PSFGKVYYID GSFSWGARAK FTCMEGYYVV GMSVLHCVLK GDDEAYWNGY PPHCEKIYCL PPPKIKNGTH TLTDINVFKY HEAVSYSCDP TPGPDKFSLV GTSMIFCAGH NTWSNSPPEC KVVKCPNPVL QNGRLISGAG EIFSYQSTVM FECLQGFYME GSSMVICSAN NSWEPSIPKC LKGPRPTHPT KPPVYNYTGY PSPREGIFSQ ELDAW
Videos
SDS
Documents
Certificates