missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CLEC4E / CLECSF9 Protein
A DNA sequence encoding the Mus musculus (Mouse) CLEC4E / CLECSF9, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_064332.1 |
---|---|
Gene ID (Entrez) | 56619 |
Name | CLEC4E / CLECSF9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022671
|
enQuireBio™
QP8099-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042671
|
enQuireBio™
QP8099-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032671
|
enQuireBio™
QP8099-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052671
|
enQuireBio™
QP8099-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062671
|
enQuireBio™
QP8099-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072671
|
enQuireBio™
QP8099-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_064332.1 | |
CLEC4E / CLECSF9 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56619 | |
Research Use Only | |
Clec4e | |
Recombinant Protein | |
E. coli | |
TYRSSQISGQ NLQPHRNIKE LSCYSEASGS VKNCCPLNWK HYQSSCYFFS TTTLTWSSSL KNCSDMGAHL VVIDTQEEQE FLFRTKPKRK EFYIGLTDQV VEGQWQWVDD TPFTESLSFW DAGEPNNIVL VEDCATIRDS SNSRKNWNDI PCFYSMPWIC EMPEISPLD | |
Greater than 90% as determined by SDS-PAGE. |