missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Complement C3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Complement C3, was expressed in the hosts and tags indicated.
300.00 € - 3260.00 €
Specifications
Accession Number | NP_033908.2 |
---|---|
Gene ID (Entrez) | 12266 |
Name | Complement C3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058670
|
enQuireBio™
QP7399-EC-10UG |
10 μg |
300.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048670
|
enQuireBio™
QP7399-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068670
|
enQuireBio™
QP7399-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078670
|
enQuireBio™
QP7399-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088670
|
enQuireBio™
QP7399-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098670
|
enQuireBio™
QP7399-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033908.2 | |
Complement C3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
12266 | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
SEETKQNEAF SLTAKGKGRG TLSVVAVYHA KLKSKVTCKK FDLRVSIRPA PETAKKPEEA KNTMFLEICT KYLGDVDATM SILDISMMTG FAPDTKDLEL LASGVDRYIS KYEMNKAFSN KNTLIIYLEK ISHTEEDCLT FKVHQYFNVG LIQPGSVKVY SYYNLEESCT RFYHPEKDDG MLSKLCHSEM CRCAEENCFM QQSQEKINLN VRLDKACEPG VDYVYKTELT NIELLDDFDE YTMTIQQVIK SGSDEVQAGQ QRKFISHIKC RNALKLQKGK KYLMWGLSSD LWGEKPNTSY IIGKDTWVEH WPEAEECQDQ KYQKQCEELG AFTESMVVYG CPN | |
Greater than 80% as determined by SDS-PAGE. |