missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Protein
Recombinant Protein
2490.00 € - 4060.00 €
Specifications
Accession Number | NP_038683.1 |
---|---|
Gene ID (Entrez) | 20315 |
Name | CXCL12 / SDF-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956159
|
enQuireBio™
QP5454-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946159
|
enQuireBio™
QP5454-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_038683.1 | |
CXCL12 / SDF-1 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse CXCL12 / SDF-1 induces Primary human T cell chemotaxis (significanty different from control at Mouse CXCL12 / SDF-1 concentrations greater than 5-50 ng/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
20315 | |
Research Use Only | |
CXCL12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK | |
Greater than 95% as determined by SDS-PAGE. |