missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Desmoglein-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Desmoglein-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5951-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_085099.2 | |
Desmoglein-3 Protein | |
Research Use Only | |
Dsg3 | |
Recombinant Protein | |
Yeast | |
EWVKFAKPCR EREDNSRRNP IAKITSDFQK NQKITYRISG VGIDQPPFGI FVVDPNNGDI NITAIVDREE TPSFLITCRA LNALGQDVER PLILTVKILD VNDNPPIFSQ TIFKGEIEEN SASNSLVMIL NATDADEPNH MNSKIAFKIV SQEPAGMSMF LISRNTGEVR TLTSSLDREQ ISSYHLVVSG ADNDGTGLST QCECSIKIKD VNDNFPVLRE SQYSARIEEN TLNAELLRFQ VTDWDEEYTD NWLAVYFFTS GNEGNWFEIE TDPRTNEGIL KVVKALDYEQ VQSMQFSIAV RNKAEFHQSV ISQYRVQSTP VTIQVIDVRE GISFRPPSKT FTVQRGVSTN KLVGYILGTY QATDEDTGKA ASSVRYVLGR NDGGLLVIDS KTAQIKFVKN IDRDSTFIVN KTISAEVLAI DENTGKTSTG TIYVEVPSFN ENCPSVVLEK KDICTSSPSV TLSVRTLDRG KYTGPYTVSL EEQPLKLPVM WTITTLNATS ALLQAQQQVS PGVYNVPVIV KDNQDGLCDT PESLTLTVCQ CDDRSMCRAP IPSREPNTYG ESSWRLGP | |
Greater than 90% as determined by SDS-PAGE. |
13512 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |