missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse E3 ubiquitin-protein ligase Mdm2 Protein
A DNA sequence encoding the Mus musculus (Mouse) E3 ubiquitin-protein ligase Mdm2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001275515.1 |
---|---|
Gene ID (Entrez) | 17246 |
Name | E3 ubiquitin-protein ligase Mdm2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022680
|
enQuireBio™
QP6353-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042680
|
enQuireBio™
QP6353-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032680
|
enQuireBio™
QP6353-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052680
|
enQuireBio™
QP6353-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062680
|
enQuireBio™
QP6353-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072680
|
enQuireBio™
QP6353-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001275515.1 | |
E3 ubiquitin-protein ligase Mdm2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17246 | |
Research Use Only | |
Mdm2 | |
Recombinant Protein | |
E. coli | |
MCNTNMSVST EGAASTSQIP ASEQETLVRP KPLLLKLLKS VGAQNDTYTM KEIIFYIGQY IMTKRLYDEK QQHIVYCSND LLGDVFGVPS FSVKEHRKIY AMIYRNLVAV SQQDSGTSLS ESRRQPEGGS DLKDPLQAPP EEKPSSSDLI SRLSTSSRRR SISETEENTD ELPGERHRKR RRSLSFDPSL GLCELREMCS GGSSSSSSSS SESTETPSHQ DLDDGVSEHS GDCLDQDSVS DQFSVEFEVE SLDSEDYSLS DEGHELSDED DEVYRVTVYQ TGESDTDSFE GDPEISLADY WKCTSCNEMN PPLPSHCKRC WTLRENWLPD DKGKDKVEIS EKAKLENSAQ AEEGLDVPDG KKLTENDAKE PCAEEDSEEK AEQTPLSQES DDYSQPSTSS SIVYSSQESV KELKEETQDK DESVESSFSL NAIEPCVICQ GRPKNGCIVH GKTGHLMSCF TCAKKLKKRN KPCPVCRQPI QMIVLTYFN | |
Greater than 90% as determined by SDS-PAGE. |