missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EIF4E Protein
A cDNA sequence encoding the EIF4E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11772-5ug
Additional Details : Weight : 0.01000kg
Specifications
EIF4E Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
EIF4E protein solution (1 mg/ml) 20mM Tris-HCl Buffer (pH 8.0), 10% glycerol, 1mM DTT, 0.1M NaCl and 0.1mM PMSF. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMATVEP ETTPTTNPPP AEEEKTESNQ EVANPEHYIK HPLQNRWALW FFKNDKSKTW QANLRLISKF DTVEDFWALY NHIQLSSNLM PGCDYSLFKD GIEPMWEDEK NKRGGRWLIT LNKQQRRSDL DRFWLETLLC LIGESFDDYS DDVCGAVVNV RAKGDKIAIW TTECENRDAV THIGRVYKER LGLPPKIVIG YQSHADTATK SGSTTKNRFV V |