missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse F3 Protein

A cDNA sequence encoding the F3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11827-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988498

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

F3 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
F3 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
ADPAGIPEKA FNLTWISTDF KTILEWQPKP TNYTYTVQIS DRSRNWKNKC FSTTDTECDL TDEIVKDVTW AYEAKVLSVP RRNSVHGDGD QLVIHGEEPP FTNAPKFLPY RDTNLGQPVI QQFEQDGRKL NVVVKDSLTL VRKNGTFLTL RQVFGKDLGY IITYRKGSST GKKTNITNTN EFSIDVEEGV SYCFFVQAMI FSRKTNQNSP GSSTVCTEQW KSFLGEHHHH HH
Videos
SDS
Documents
Certificates