missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fas apoptotic inhibitory molecule 3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Fas apoptotic inhibitory molecule 3, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_081252.1 |
---|---|
Gene ID (Entrez) | 69169 |
Name | Fas apoptotic inhibitory molecule 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929389
|
enQuireBio™
QP6017-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949389
|
enQuireBio™
QP6017-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939389
|
enQuireBio™
QP6017-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959389
|
enQuireBio™
QP6017-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969389
|
enQuireBio™
QP6017-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979389
|
enQuireBio™
QP6017-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_081252.1 | |
Fas apoptotic inhibitory molecule 3 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
69169 | |
Research Use Only | |
Faim3 | |
Recombinant Protein | |
E. coli | |
RVLPEVQLNV EWGGSIIIEC PLPQLHVRMY LCRQMAKPGI CSTVVSNTFV KKEYERRVTL TPCLDKKLFL VEMTQLTEND DGIYACGVGM KTDKGKTQKI TLNVHNEYPE PFWEDEWTSE RPRWLHRFLQ HQMPWLHGSE HPSSSGVIAK VTTPAPKTEA PPVHQPSSIT SVTQHPRVYR AFSVSATKSP ALLPATTASK TSTQQAIRPL EASYSHHTRL HEQRTRHHGP HYGREDRGLH IPIPE | |
Greater than 90% as determined by SDS-PAGE. |