missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fas apoptotic inhibitory molecule 3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Fas apoptotic inhibitory molecule 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6017-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929389

  • 680.00 € / 100µg
Estimated Shipment: 23-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_081252.1
Fas apoptotic inhibitory molecule 3 Protein
100 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
69169
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Faim3
Recombinant Protein
E. coli
RVLPEVQLNV EWGGSIIIEC PLPQLHVRMY LCRQMAKPGI CSTVVSNTFV KKEYERRVTL TPCLDKKLFL VEMTQLTEND DGIYACGVGM KTDKGKTQKI TLNVHNEYPE PFWEDEWTSE RPRWLHRFLQ HQMPWLHGSE HPSSSGVIAK VTTPAPKTEA PPVHQPSSIT SVTQHPRVYR AFSVSATKSP ALLPATTASK TSTQQAIRPL EASYSHHTRL HEQRTRHHGP HYGREDRGLH IPIPE
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers