missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Frataxin, mitochondrial Protein

A DNA sequence encoding the Mus musculus (Mouse) Frataxin, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6059-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989679

Quantity Price
1 2825.92 € / 1mg
3 2658.91 € / 1mg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

NP_032070.1
Frataxin, mitochondrial Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
14297
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Fxn
Recombinant Protein
E. coli
LGTLDNPSSL DETAYERLAE ETLDSLAEFF EDLADKPYTL EDYDVSFGDG VLTIKLGGDL GTYVINKQTP NKQIWLSSPS SGPKRYDWTG KNWVYSHDGV SLHELLAREL TKALNTKLDL SSLAYSGKGT
Videos
SDS
Documents
Certificates