missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GAD65 / GAD2 Protein
A DNA sequence encoding the Mus musculus (Mouse) GAD65 / GAD2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_032104.2 |
---|---|
Gene ID (Entrez) | 14417 |
Name | GAD65 / GAD2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909709
|
enQuireBio™
QP6062-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929709
|
enQuireBio™
QP6062-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919709
|
enQuireBio™
QP6062-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939709
|
enQuireBio™
QP6062-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949709
|
enQuireBio™
QP6062-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959709
|
enQuireBio™
QP6062-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_032104.2 | |
GAD65 / GAD2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
14417 | |
Research Use Only | |
Gad2 | |
Recombinant Protein | |
E. coli | |
MASPGSGFWS FGSEDGSADP ENPGTARAWC QVAQKFTGGI GNKLCALLYG DSGKPAEGGG SVTSRAATGK VACTCDQKPC NCPKGDVNYA FLHATDLLPA CDGERPTLAF LQDVMNILLQ YVVKSFDRST KVIDFHYPNE LLQEYNWELA DQPQNLEEIL THCQTTLKYA IKTGHPRYFN QLSTGLDMVG LAADWLTSTA NTNMFTYEIA PVFVLLEYVT LKKMREIIGW PGGSGDGIFS PGGAISNMYA MLIARYKMFP EVKEKGMAAV PRLIAFTSEH SHFSLKKGAA ALGIGTDSVI LIKCDERGKM IPSDLERRIL EVKQKGFVPF LVSATAGTTV YGAFDPLLAV ADICKKYKIW MHVDAAWGGG LLMSRKHKWK LSGVERANSV TWNPHKMMGV PLQCSALLVR EEGLMQSCNQ MHASYLFQQD KHYDLSYDTG DKALQCGRHV DVFKLWLMWR AKGTTGFEAH IDKCLELAEY LYTIIKNREG YEMVFDGKPQ HTNVCFWFVP PSLRTLEDNE ERMSRLSKVA PVIKARMMEY GTTMVSYQPL GDKVNFFRMV ISNPAATHQD IDFLIEEIER LGQDL | |
Greater than 90% as determined by SDS-PAGE. |