missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GDF15 Protein
A cDNA sequence encoding the GDF15 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11948-5ug
Additional Details : Weight : 0.01000kg
Specifications
23886 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSAHAHPR DSCPLGPGRC CHLETVQATL EDLGWSDWVL SPRQLQLSMC VGECPHLYRS ANTHAQIKAR LHGLQPDKVP APCCVPSSYT PVVLMHRTDS GVSLQTYDDL VARGCHCA | |
Greater than 90.0% as determined by SDS-PAGE. |
GDF15 Protein | |
Research Use Only | |
Gdf15 | |
Mouse | |
His | |
GDF15 protein solution (1.0 mg/ml) containing 20mM Phosphate buffer (pH 8.0) and 10% glycerol. |