missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GDNF Protein

Recombinant Protein

Brand:  enQuireBio™ QP5446-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986139

  • 6169.88 € / 1mg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001288261.1
GDNF Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Bioactivity of recombinant GDNF is confirmed by C6 cell proliferation assay (see figure).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
14573
Greater than 95% as determined by SDS-PAGE.
Research Use Only
GDNF
Recombinant Protein
E. coli
MSPDKQAALP RRENRNRQAA AASPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCESAETMY DKILKNLSRS RRLTSDKVGQ ACCRPVAFDD DLSFLDDNLV YHILRKHSAK RCGCI
Videos
SDS
Documents
Certificates