missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glucagon receptor Protein
A DNA sequence encoding the Mus musculus (Mouse) Glucagon receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6073-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032127.2 | |
Glucagon receptor Protein | |
Research Use Only | |
Gcgr | |
Recombinant Protein | |
E. coli | |
AQVMDFLFEK WKLYSDQCHH NLSLLPPPTE LVCNRTFDKY SCWPDTPPNT TANISCPWYL PWYHKVQHRL VFKRCGPDGQ WVRGPRGQPW RNASQCQLDD EEIEVQKGVA KMYSSQQ | |
Greater than 90% as determined by SDS-PAGE. |
14527 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |