missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GMFB Protein
A cDNA sequence encoding the GMFB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12003-1mg
Specifications
GMFB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SESLVVCDVA EDLVEKLRKF RFRKETHNAA IIMKIDKDER LVVLDEELEGVSPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQMMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The GMF-beta protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |