missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GSTP2 Protein

A cDNA sequence encoding the GSTP2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12063-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989238

  • 3401.86 € / 1mg

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

14869
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Gstp2
Mouse
Untagged
GSTP2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
GSTP2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPPYTIV YFPSPGRCEA MRMLLADQGQ SWKEEVVTID TWMQGLLKPT CLYGQLPKFE DGDLTLYQSN AILRHLGRSL GLYGKNQREA AQVDMVNDGV EDLRGKYGTM IYRNYENGKN DYVKALPGHL KPFETLLSQN QGGKAFIVGD QISFADYNLL DLLLIHQVLA PGCLDNFPLL SAYVARLSAR PKIKAFLSSP EHVNRPINGN GKQ
Videos
SDS
Documents
Certificates