missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Histone H2B type 1-M Protein
A DNA sequence encoding the Mus musculus (Mouse) Histone H2B type 1-M, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6160-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_835507.1 | |
Histone H2B type 1-M Protein | |
Research Use Only | |
Hist1h2bm | |
Recombinant Protein | |
E. coli | |
PEPTKSAPAP KKGSKKAVTK AQKKDGKKRK RSRKESYSVY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSSK | |
Greater than 90% as determined by SDS-PAGE. |
319186 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |