missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse HSP70 / HSPA1A Protein
A DNA sequence encoding the Mus musculus (Mouse) HSP70 / HSPA1A, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_034608.2 |
---|---|
Gene ID (Entrez) | 15511 |
Name | HSP70 / HSPA1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031410
|
enQuireBio™
QP6185-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051410
|
enQuireBio™
QP6185-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041410
|
enQuireBio™
QP6185-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061410
|
enQuireBio™
QP6185-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071410
|
enQuireBio™
QP6185-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081410
|
enQuireBio™
QP6185-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034608.2 | |
HSP70 / HSPA1A Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
15511 | |
Research Use Only | |
Hspa1b | |
Recombinant Protein | |
E. coli | |
AKNTAIGIDL GTTYSCVGVF QHGKVEIIAN DQGNRTTPSY VAFTDTERLI GDAAKNQVAL NPQNTVFDAK RLIGRKFGDA VVQSDMKHWP FQVVNDGDKP KVQVNYKGES RSFFPEEISS MVLTKMKEIA EAYLGHPVTN AVITVPAYFN DSQRQATKDA GVIAGLNVLR IINEPTAAAI AYGLDRTGKG ERNVLIFDLG GGTFDVSILT IDDGIFEVKA TAGDTHLGGE DFDNRLVSHF VEEFKRKHKK DISQNKRAVR RLRTACERAK RTLSSSTQAS LEIDSLFEGI DFYTSITRAR FEELCSDLFR GTLEPVEKAL RDAKMDKAQI HDLVLVGGST RIPKVQKLLQ DFFNGRDLNK SINPDEAVAY GAAVQAAILM GDKSENVQDL LLLDVAPLSL GLETAGGVMT ALIKRNSTIP TKQTQTFTTY SDNQPGVLIQ VYEGERAMTR DNNLLGRFEL SGIPPAPRGV PQIEVTFDID ANGILNVTAT DKSTGKANKI TITNDKGRLS KEEIERMVQE AERYKAEDEV QRDRVAAKNA LESYAFNMKS AVEDEGLKGK LSEADKKKVL DKCQEVISWL DSNTLADKEE FVHKREELER VCSPIISGLY QGAGAPGAGG FGAQAPPKGA SGSGPTIEEV D | |
Greater than 90% as determined by SDS-PAGE. |