missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse I-TAC / CXCL11 Protein

A DNA sequence encoding the Mus musculus (Mouse) I-TAC / CXCL11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8656-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045801

  • 3375.00 € / 1mg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_062367.1
I-TAC / CXCL11 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
56066
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CXCL11
Recombinant Protein
E. coli
FLMFKQGRCL CIGPGMKAVK MAEIEKASVI YPSNGCDKVE VIVTMKAHKR QRCLDPRSKQ ARLIMQAIEK KNFLRRQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers