missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IDO1 / IDO Protein
A DNA sequence encoding the Mus musculus (Mouse) IDO1 / IDO, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_001280619.1 |
---|---|
Gene ID (Entrez) | 15930 |
Name | IDO1 / IDO Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037821
|
enQuireBio™
QP9192-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077821
|
enQuireBio™
QP9192-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047821
|
enQuireBio™
QP9192-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067821
|
enQuireBio™
QP9192-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087821
|
enQuireBio™
QP9192-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057821
|
enQuireBio™
QP9192-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001280619.1 | |
IDO1 / IDO Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
15930 | |
Research Use Only | |
IDO1 | |
Recombinant Protein | |
Yeast | |
MALSKISPTE GSRRILEDHH IDEDVGFALP HPLVELPDAY SPWVLVARNL PVLIENGQLR EEVEKLPTLS TDGLRGHRLQ RLAHLALGYI TMAYVWNRGD DDVRKVLPRN IAVPYCELSE KLGLPPILSY ADCVLANWKK KDPNGPMTYE NMDILFSFPG GDCDKGFFLV SLLVEIAASP AIKAIPTVSS AVERQDLKAL EKALHDIATS LEKAKEIFKR MRDFVDPDTF FHVLRIYLSG WKCSSKLPEG LLYEGVWDTP KMFSGGSAGQ SSIFQSLDVL LGIKHEAGKE SPAEFLQEMR EYMPPAHRNF LFFLESAPPV REFVISRHNE DLTKAYNECV NGLVSVRKFH LAIVDTYIMK PSKKKPTDGD KSEEPSNVES RGTGGTNPMT FLRSVKDTTE KALLSWP | |
Greater than 90% as determined by SDS-PAGE. |