missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IgG Protein

A cDNA sequence encoding the IgG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12386-HIS-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990249

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

IgG Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
IgG protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
ADPEPRGPTI KPCPPCKCPA PNLLGGPSVF IFPPKIKDVL MISLSPIVTC VVVDVSEDDP DVQISWFVNN VEVHTAQTQT HREDYNSTLR VVSALPIQHQ DWMSGKEFKC KVNNKDLPAP IERTISKPKG SVRAPQVYVL PPPEEEMTKK QVTLTCMVTD FMPEDIYVEW TNNGKTELNY KNTEPVLDSD GSYFMYSKLR VEKKNWVERN SYSCSVVHEG LHNHHTTKSF SRTPGKHHHH HH
Videos
SDS
Documents
Certificates