missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Ihh Protein
Recombinant Protein
107.00 € - 3770.00 €
Specifications
Accession Number | NP_034674.2 |
---|---|
Gene ID (Entrez) | 16147 |
Name | Ihh Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15996219
|
enQuireBio™
QP5480-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976219
|
enQuireBio™
QP5480-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956219
|
enQuireBio™
QP5480-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986219
|
enQuireBio™
QP5480-500UG |
500 μg |
2320.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966219
|
enQuireBio™
QP5480-1MG |
1 mg |
3770.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_034674.2 | |
Ihh Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse Ihh induced alkaline phosphatase activity in CCL-226 cells (typical ED50 is ? 10 ug/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
16147 | |
Research Use Only | |
Ihh | |
Recombinant Protein | |
E. coli | |
MIIGPGRVVG . SRRRPPRKL V. PLAYKQFS PN. VPEKTLG ASG. RYEGKI ARSS. ERFKE LTPNY. NPDI IFKDEE. NTG ADRLMTQRCK DRLNSLA. IS VMNQWPGV. K LRVTEGWDE. DGHHSEESLH . YEGRAVDIT T. SDRDRNKY GL. LARLAVE AGF. DWVYYE SKAH. VHCSV KSEHSAAAKT GG | |
Greater than 95% as determined by SDS-PAGE. |