missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-11 / interleukin 11 Protein
Recombinant Protein
Brand: enQuireBio™ QP5472-1mg
Specifications
NP_032376.1 | |
IL-11 / interleukin 11 Protein | |
Research Use Only | |
IL11 | |
Recombinant Protein | |
E. coli | |
MPGPPAGSPR VSSDPRADLD SAVLLTRSLL ADTRQLAAQM RDKFPADGDH SLDSLPTLAM SAGTLGSLQL PGVLTRLRVD LMSYLRHVQW LRRAGGPSLK TLEPELGALQ ARLERLLRRL QLLMSRLALP QAAPDQPVIP LGPPASAWGS IRAAHAILGG LHLTLDWAVR GLLLLKTRL | |
Greater than 95% as determined by SDS-PAGE. |
16156 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 2.5 ng/mL in the T11 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^5 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |