missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-12AB Protein
Recombinant Protein
107.00 € - 10580.00 €
Specifications
Accession Number | NP_001290173.1 |
---|---|
Gene ID (Entrez) | 16160 |
Name | IL-12AB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15915669
|
enQuireBio™
QP5204-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995659
|
enQuireBio™
QP5204-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985659
|
enQuireBio™
QP5204-100UG |
100 μg |
1885.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925669
|
enQuireBio™
QP5204-500UG |
500 μg |
6400.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905669
|
enQuireBio™
QP5204-1MG |
1 mg |
10580.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001290173.1 | |
IL-12AB Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ng/mL in the Mouse primary T cell induced expression of CD25 assay. The bioactivity acceptance criteria is set at 5 x 10^4 units/mg. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
16160 | |
Research Use Only | |
IL12 | |
Recombinant Protein | |
Human (HEK293) | |
p35:MRVIPV SGPARCLSQS RNLLKTTDDM VKTAREKLKH YSCTAEDIDH EDITRDQTST LKTCLPLELH KNESCLATRE TSSTTRGSCL PPQKTSLMMT LCLGSIYEDL KMYQTEFQAI NAALQNHNHQ QIILDKGMLV AIDELMQSLN HNGETLRQKP PVGEADPYRV KMKLCILLHA FSTRVVTINR VMGYLSSA p40:MWELEKDV YVVEVDWTPD APGETVNLTC DTPEEDDITW TSDQRHGVIG SGKTLTITVK EFLDAGQYTC HKGGETLSHS HLLLHKKENG IWSTEILKNF KNKTFLKCEA PNYSGRFTCS WLVQRNMDLK FNIKSSSSSP DSRAVTCGMA SLSAEKVTLD QRDYEKYSVS CQEDVTCPTA EETLPIELAL EARQQNKYEN YSTSFFIRDI IKPDPPKNLQ MKPLKNSQVE VSWEYPDSWS TPHSYFSLKF FVRIQRKKEK MKETEEGCNQ KGAFLVEKTS TEVQCKGGNV CVQAQDRYYN SSCSKWACVP CRVRS | |
Greater than 95% as determined by SDS-PAGE. |