Learn More
enQuireBio™ Recombinant Mouse IL-15 Protein
Recombinant Protein
Brand:  enQuireBio™ QP5406-2ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15986059
Quantity Price
1 89.91 € / 2µg
10 84.58 € / 2µg
Estimated Shipment: 18-11-2021
Specifications
NP_001241676.1
IL-15 Protein
2 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity in the CTLL-2 cell proliferation assay.
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
16168
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL15
Recombinant Protein
E. coli
MNWIDVRYDL EKIESLIQSI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VAESGCKECE ELEEKTFTEF LQSFIRIVQM FINTS