missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL-15 Protein
Recombinant Protein
107.00 € - 5365.00 €
Specifications
Accession Number | NP_001241676.1 |
---|---|
Gene ID (Entrez) | 16168 |
Name | IL-15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986059
|
enQuireBio™
QP5406-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966059
|
enQuireBio™
QP5406-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956059
|
enQuireBio™
QP5406-100UG |
100 μg |
700.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996059
|
enQuireBio™
QP5406-500UG |
500 μg |
3285.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976059
|
enQuireBio™
QP5406-1MG |
1 mg |
5365.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001241676.1 | |
IL-15 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the CTLL-2 cell proliferation assay. | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
16168 | |
Research Use Only | |
IL15 | |
Recombinant Protein | |
E. coli | |
MNWIDVRYDL EKIESLIQSI HIDTTLYTDS DFHPSCKVTA MNCFLLELQV ILHEYSNMTL NETVRNVLYL ANSTLSSNKN VAESGCKECE ELEEKTFTEF LQSFIRIVQM FINTS | |
Greater than 95% as determined by SDS-PAGE. |