missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL 28A Protein
A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 330496 |
---|---|
Name | IL 28A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Ifnl2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954928
|
enQuireBio™
QP10708-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944928
|
enQuireBio™
QP10708-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934928
|
enQuireBio™
QP10708-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
330496 | |
Research Use Only | |
Ifnl2 | |
Recombinant Protein | |
E. coli | |
DPVPRATRLP VEAKDCHIAQ FKSLSPKELQ AFKKAKDAIE KRLLEKDMRC SSHLISRAWD LKQLQVQERP KALQAEVALT LKVWENMTDS ALATILGQPL HTLSHIHSQL QTCTQLQATA EPKPPSRRLS RWLHRLQEAQ SKETPGCLED SVTSNLFRLL TRDLKCVASG DQCV | |
Greater than 95% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 28A Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis(EMC) virus is less than 3 ng/ml, corresponding to a specific activity of > 3.3 M 105 IU/mg. | |
Mouse | |
Untagged | |
The Mouse IL-28A is lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4 and 5% trehalose. |