missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL17 / IL17A Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_034682.1 |
---|---|
Gene ID (Entrez) | 16171 |
Name | IL17 / IL17A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986189
|
enQuireBio™
QP5466-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966189
|
enQuireBio™
QP5466-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976189
|
enQuireBio™
QP5466-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956189
|
enQuireBio™
QP5466-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_034682.1 | |
IL17 / IL17A Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the Production of IL-6 by mouse 3T3 cells assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
16171 | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
E. coli | |
MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA | |
Greater than 95% as determined by SDS-PAGE. |