missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL22 / Interleukin 22 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5485-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986229

  • 3755.49 € / 500µg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_058667.1
IL22 / Interleukin 22 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 1 ng/mL in the Production of IL-10 from human COLO-205 cells assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg.
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
50929
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL22
Recombinant Protein
E. coli
MLPVNTRCK LEVSNFQQPY IVNRTFMLAK EASLADNNTD VRLIGEKLFR GVSAKDQCYL MKQVLNFTLE DVLLPQSDRF QPYMQEVVPF LTKLSNQLSS CHISGDDQNI QKNVRRLKET VKKLGESGEI KAIGELDLLF MSLRNACV
Videos
SDS
Documents
Certificates