missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL22 / Interleukin 22 Protein
Recombinant Protein
4325.00 € - 7105.00 €
Specifications
Accession Number | NP_058667.1 |
---|---|
Gene ID (Entrez) | 50929 |
Name | IL22 / Interleukin 22 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986229
|
enQuireBio™
QP5485-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976229
|
enQuireBio™
QP5485-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_058667.1 | |
IL22 / Interleukin 22 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the Production of IL-10 from human COLO-205 cells assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
50929 | |
Research Use Only | |
IL22 | |
Recombinant Protein | |
E. coli | |
MLPVNTRCK LEVSNFQQPY IVNRTFMLAK EASLADNNTD VRLIGEKLFR GVSAKDQCYL MKQVLNFTLE DVLLPQSDRF QPYMQEVVPF LTKLSNQLSS CHISGDDQNI QKNVRRLKET VKKLGESGEI KAIGELDLLF MSLRNACV | |
Greater than 95% as determined by SDS-PAGE. |