missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL34 Protein
A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12422-2ug
Additional Details : Weight : 0.01000kg
Specifications
76527 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNENLEIWTL TQDKECDLTG YLRGKLQYKN RLQYMKHYFP INYRIAVPYE GVLRVANITR LQKAHVSERE LRYLWVLVSL NATESVMDVL LEGHPSWKYL QEVQTLLENV QRSLMDVEIG PHVEAVLSLL STPGLSLKLV RPKALLDNCF RVMELLYCSC CKQSPILKWQ DCELPRLHPH SPGSLMQCTA TNVYPLSRQT PTSLPGSPSS SHGSLP | |
Greater than 85% as determined by SDS-PAGE. |
IL34 Protein | |
Research Use Only | |
Il34 | |
Mouse | |
His | |
IL34 protein solution (0.5 mg/ml) in 20mM Tris-HCl buffer (pH 8.0) and 10% Glycerol. |