missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL5RA Protein
A cDNA sequence encoding the IL5RA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12426-10ug
Additional Details : Weight : 0.01000kg
Specifications
16192 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DLLNHKKFLL LPPVNFTIKA TGLAQVLLHW DPNPDQEQRH VDLEYHVKIN APQEDEYDTR KTESKCVTPL HEGFAASVRT ILKSSHTTLA SSWVSAELKA PPGSPGTSVT NLTCTTHTVV SSHTHLRPYQ VSLRCTWLVG KDAPEDTQYF LYYRFGVLTE KCQEYSRDAL NRNTACWFPR TFINSKGFEQ LAVHINGSSK RAAIKPFDQL FSPLAIDQVN PPRNVTVEIE SNSLYIQWEK PLSAFPDHCF NYELKIYNTK NGHIQKEKLI ANKFISKIDD VSTYSIQVRA AVSSPCRMPG RWGEWSQPIY VGKERKSLVE WHLEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
IL5RA Protein | |
Research Use Only | |
Il5ra | |
Mouse | |
His | |
IL5RA protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |