missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IL6R Protein
A cDNA sequence encoding the IL6R was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 16195 |
---|---|
Name | IL6R Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Il6st |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970389
|
enQuireBio™
QP12428-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950389
|
enQuireBio™
QP12428-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960389
|
enQuireBio™
QP12428-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
16195 | |
Research Use Only | |
Il6st | |
Mouse | |
His | |
IL6R protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |
IL6R Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
QLLEPCGYIY PEFPVVQRGS NFTAICVLKE ACLQHYYVNA SYIVWKTNHA AVPREQVTVI NRTTSSVTFT DVVLPSVQLT CNILSFGQIE QNVYGVTMLS GFPPDKPTNL TCIVNEGKNM LCQWDPGRET YLETNYTLKS EWATEKFPDC QSKHGTSCMV SYMPTYYVNI EVWVEAENAL GKVSSESINF DPVDKVKPTP PYNLSVTNSE ELSSILKLSW VSSGLGGLLD LKSDIQYRTK DASTWIQVPL EDTMSPRTSF TVQDLKPFTE YVFRIRSIKD SGKGYWSDWS EEASGTTYED RPSRPPSFWY KTNPSHGQEY RSVRLIWKAL PLSEANGKIL DYEVILTQSK SVSQTYTVTG TELTVNLTND RYVASLAARN KVGKSAAAVL TIPSPHVTAA YSVVNLKAFP KDNLLWVEWT PPPKPVSKYI LEWCVLSENA PCVEDWQQED ATVNRTHLRG RLLESKCYQI TVTPVFATGP GGSESLKAYL KQAAPARGPT VRTKKVGKNE AVLAWDQIPV DDQNGFIRNY SISYRTSVGK EMVVHVDSSH TEYTLSSLSS DTLYMVRMAA YTDEGGKDGP EFTFTTPKFA QGEIELEHHH HHH | |
Greater than 95.0% as determined by SDS-PAGE. |