missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Interferon regulatory factor 5 Protein

A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6234-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031800

  • 735.00 € / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001239311.1
Interferon regulatory factor 5 Protein
Research Use Only
Irf5
Recombinant Protein
E. coli
MNHSAPGIPP PPRRVRLKPW LVAQVNSCQY PGLQWVNGEK KLFYIPWRHA TRHGPSQDGD NTIFKAWAKE TGKYTEGVDE ADPAKWKANL RCALNKSRDF QLFYDGPRDM PPQPYKIYEV CSNGPAPTES QPTDDYVLGE EEEEEEEELQ RMLPGLSITE PALPGPPNAP YSLPKEDTKW PPALQPPVGL GPPVPDPNLL APPSGNPAGF RQLLPEVLEP GPLASSQPPT EPLLPDLLIS PHMLPLTDLE IKFQYRGRAP RTLTISNPQG CRLFYSQLEA TQEQVELFGP VTLEQVRFPS PEDIPSDKQR FYTNQLLDVL DRGLILQLQG QDLYAIRLCQ CKVFWSGPCA LAHGSCPNPI QREVKTKLFS LEQFLNELIL FQKGQTNTPP PFEIFFCFGE EWPDVKPREK KLITVQVVPV AARLLLEMFS GELSWSADSI RLQISNPDLK DHMVEQFKEL HHLWQSQQQL QPMVQAPPVA GLDASQGPWP MHPVGMQ
Greater than 90% as determined by SDS-PAGE.
27056
100 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers