missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Kallikrein 1-related peptidase b22 Protein
A DNA sequence encoding the Mus musculus (Mouse) Kallikrein 1-related peptidase b22, was expressed in the hosts and tags indicated.
348.00 € - 3590.00 €
Specifications
Accession Number | NP_034244.1 |
---|---|
Gene ID (Entrez) | 13646 |
Name | Kallikrein 1-related peptidase b22 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027390
|
enQuireBio™
QP7102-10UG |
10 μg |
348.00 €
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067390
|
enQuireBio™
QP7102-50UG |
50 μg |
493.00 €
50µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017390
|
enQuireBio™
QP7102-100UG |
100 μg |
820.00 €
100µg |
Estimated Shipment: 09-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047390
|
enQuireBio™
QP7102-200UG |
200 μg |
1330.00 €
200µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057390
|
enQuireBio™
QP7102-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037390
|
enQuireBio™
QP7102-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_034244.1 | |
Kallikrein 1-related peptidase b22 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
13646 | |
Research Use Only | |
Klk1b22 | |
Recombinant Protein | |
E. coli | |
ILGGFKCEKN SQPWQVAVYY LDEYLCGGVL LDRNWVLTAA HCYEDKYNIW LGKNKLFQDE PSAQHRLVSK SFPHPDFNMS LLQSVPTGAD LSNDLMLLRL SKPADITDVV KPIDLPTTEP KLGSTCLASG WGSINQLIYQ NPNDLQCVSI KLHPNEVCVK AHILKVTDVM LCAGEMNGGK DTCKGDSGGP LICDGVLQGI TSWGSTPCGE PNAPAIYTKL IKFTSWIKDT MAKNP | |
Greater than 90% as determined by SDS-PAGE. |