missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse KGF 2 Protein
A cDNA sequence encoding the KGF 2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10735-25ug
Additional Details : Weight : 0.01000kg
Specifications
KGF 2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
QALGQDMVSQ EATNCSSSSS SFSSPSSAGR HVRSYNHLQG DVRWRRLFSF TKYFLTIEKN GKVSGTKNED CPYSVLEITS VEIGVVAVKA INSNYYLAMN KKGKLYGSKE FNNDCKLKER IEENGYNTYA SFNWQHNGRQ MYVALNGKGA PRRGQKTRRK NTSAHFLPMT IQT | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
25 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 containing 5% trehalose. |