missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse LAIR1 Protein

A cDNA sequence encoding the LAIR1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12525-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960709

  • 131.00 € / 2µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

LAIR1 Protein
Research Use Only
Recombinant Protein
Baculovirus
ADPQEGSLPD ITIFPNSSLM ISQGTFVTVV CSYSDKHDLY NMVRLEKDGS TFMEKSTEPY KTEDEFEIGP VNETITGHYS CIYSKGITWS ERSKTLELKV IKENVIQTPA PGPTSDTSWL KTYSIYHHHH HH
Greater than 90.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
LAIR1 protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers