missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse LGALS3 Protein

A cDNA sequence encoding the LGALS3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12563-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15990819

  • 6549.36 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LGALS3 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
The LGALS3 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol, 1mM DTT and 2mM EDTA.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMADSFSL NDALAGSGNP NPQGYPGAWG NQPGAGGYPG AAYPGAYPGQ APPGAYPGQA PPGAYPGQAP PSAYPGPTAP GAYPGPTAPG AYPGSTAPGA FPGQPGAPGA YPSAPGGYPA AGPYGVPAGP LTVPYDLPLP GGVMPRMLIT IMGTVKPNAN RIVLDFRRGN DVAFHFNPRF NENNRRVIVC NTKQDNNWGK EERQSAFPFE SGKPFKIQVL VEADHFKVAV NDAHLLQYNH RMKNLREISQ LGISGDITLT SANHAMI
Videos
SDS
Documents
Certificates