missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Lymphocyte antigen 6E Protein
A DNA sequence encoding the Mus musculus (Mouse) Lymphocyte antigen 6E, was expressed in the hosts and tags indicated.
245.00 € - 2645.00 €
Specifications
Accession Number | NP_001157508.1 |
---|---|
Gene ID (Entrez) | 17069 |
Name | Lymphocyte antigen 6E Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008751
|
enQuireBio™
QP9800-YE-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048751
|
enQuireBio™
QP9800-YE-500UG |
500 μg |
2100.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018751
|
enQuireBio™
QP9800-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038751
|
enQuireBio™
QP9800-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058751
|
enQuireBio™
QP9800-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028751
|
enQuireBio™
QP9800-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001157508.1 | |
Lymphocyte antigen 6E Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
17069 | |
Research Use Only | |
LY6E | |
Recombinant Protein | |
Yeast | |
LMCFSCTDQK NNINCLWPVS CQEKDHYCIT LSAAAGFGNV NLGYTLNKGC SPICPSENVN LNLGVASVNS YCCQSSFCNF SA | |
Greater than 90% as determined by SDS-PAGE. |