missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MCP 1 Protein

A cDNA sequence encoding the MCP 1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12663-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991149

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

20296
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Ccl2
Mouse
His
MCP-1 protein solution (0.25 mg/ml) containing PBS (pH 7.4) and 10% glycerol.
MCP 1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQPDAVNAPL TCCYSFTSKM IPMSRLESYK RITSSRCPKE AVVFVTKLKR EVCADPKKEW VQTYIKNLDR NQMRSEPTTL FKTASALRSS APLNVKLTRK SEANASTTFS TTTSSTSVGV TSVTVN
Videos
SDS
Documents
Certificates