missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MCP 1 Protein
A cDNA sequence encoding the MCP 1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6525.00 €
Specifications
Gene ID (Entrez) | 20296 |
---|---|
Name | MCP 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Ccl2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15901159
|
enQuireBio™
QP12663-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991149
|
enQuireBio™
QP12663-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981149
|
enQuireBio™
QP12663-1MG |
1 mg |
6525.00 €
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
20296 | |
Research Use Only | |
Ccl2 | |
Mouse | |
His | |
MCP-1 protein solution (0.25 mg/ml) containing PBS (pH 7.4) and 10% glycerol. |
MCP 1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQPDAVNAPL TCCYSFTSKM IPMSRLESYK RITSSRCPKE AVVFVTKLKR EVCADPKKEW VQTYIKNLDR NQMRSEPTTL FKTASALRSS APLNVKLTRK SEANASTTFS TTTSSTSVGV TSVTVN | |
Greater than 90.0% as determined by SDS-PAGE. |