missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MMP-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) MMP-3 Protein, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_034939.1 |
---|---|
Gene ID (Entrez) | 17392 |
Name | MMP-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037081
|
enQuireBio™
QP8900-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057081
|
enQuireBio™
QP8900-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047081
|
enQuireBio™
QP8900-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067081
|
enQuireBio™
QP8900-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16077081
|
enQuireBio™
QP8900-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087081
|
enQuireBio™
QP8900-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034939.1 | |
MMP-3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17392 | |
Research Use Only | |
MMP3 | |
Recombinant Protein | |
E. coli | |
PGSPKWRKSH ITYRIVNYTP DLPRQSVDSA IEKALKVWEE VTPLTFSRIS EGEADIMISF AVGEHGDFVP FDGPGTVLAH AYAPGPGING DAHFDDDERW TEDVTGTNLF LVAAHELGHS LGLYHSAKAE ALMYPVYKSS TDLSRFHLSQ DDVDGIQSLY GTPTASPDVL VVPTKSNSLE PETSPMCSST LFFDAVSTLR GEVLFFKDRH FWRKSLRTPE PEFYLISSFW PSLPSNMDAA YEVTNRDTVF IFKGNQFWAI RGHEELAGYP KSIHTLGLPA TVKKIDAAIS NKEKRKTYFF VEDKYWRFDE KKQSMEPGFP RKIAEDFPGV DSRVDAVFEA FGFLYFFSGS SQLEFDPNAK KVTHILKSNS WFNC | |
Greater than 90% as determined by SDS-PAGE. |