missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse OSM Protein
A cDNA sequence encoding the OSM was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10801-1mg
Specifications
OSM Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
NRGCSNSSSQ LLSQLQNQAN LTGNTESLLE PYIRLQNLNT PDLRAACTQH SVAFPSEDTL RQLSKPHFLS TVYTTLDRVL YQLDALRQKF LKTPAFPKLD SARHNILGIR NNVFCMARLL NHSLEIPEPT QTDSGASRST TTPDVFNTKI GSCGFLWGYH RFMGSVGRVF REWDDGSTRS R | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
OSM protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |