missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse P4HB Protein
A cDNA sequence encoding the P4HB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12948-1mg
Specifications
18453 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DALEEEDNVL VLKKSNFEEA LAAHKYLLVE FYAPWCGHCK ALAPEYAKAA AKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFKN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLSDTAAAE SLVDSSEVTV IGFFKDVESD SAKQFLLAAE AIDDIPFGIT SNSGVFSKYQ LDKDGVVLFK KFDEGRNNFE GEITKEKLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLSSFKR AAEGFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESDELTAE KITEFCHRFL EGKIKPHLMS QEVPEDWDKQ PVKVLVGANF EEVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIIIAKMDS TANEVEAVKV HSFPTLKFFP ASADRTVIDY NGERTLDGFK KFLESGGQDG AGDDEDLDLE EALEPDMEED DDQKAVKDEL LEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
P4HB Protein | |
Research Use Only | |
P4hb | |
Mouse | |
His | |
P4HB protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |