missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Phosphatidylcholine translocator ABCB4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Phosphatidylcholine translocator ABCB4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5601-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_032856.2 | |
Phosphatidylcholine translocator ABCB4 Protein | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Mouse | |
GST | |
Tris-based buffer, 50% glycerol |
18670 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Abcb4 | |
Recombinant Protein | |
E. coli | |
DAFANARGAA YVIFDIIDNN PKIDSFSERG HKPDNIKGNL EFSDVHFSYP SRANIKILKG LNLKVKSGQT VALVGNSGCG KSTTVQLLQR LYDPTEGKIS IDGQDIRNFN VRCLREIIGV VSQEPVLFST TIAENIRYGR GNVTMDEIEK AVKEANAYDF IMKLPQKFDT LVGDRGAQLS GGQKQRIAIA RALVRNPKIL LLDEATSALD TESEAEVQAA LDKAREGRTT IVIAHRLSTI RNADVIAGFE DGVIVEQGSH SELMKKEGIY FRLVNMQTAG SQILSEEFEV ELSDEKAAGD VAPNGWKARI FRNSTKKSLK SPHQNRLDEE TNELDANVPP VSFLKVLKLN KTEWPYF |