missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Small proline-rich protein 2A Protein
A DNA sequence encoding the Mus musculus (Mouse) Small proline-rich protein 2A, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_001158259.1 |
---|---|
Gene ID (Entrez) | 100042514 |
Name | Small proline-rich protein 2A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035390
|
enQuireBio™
QP6723-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055390
|
enQuireBio™
QP6723-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045390
|
enQuireBio™
QP6723-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065390
|
enQuireBio™
QP6723-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075390
|
enQuireBio™
QP6723-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085390
|
enQuireBio™
QP6723-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001158259.1 | |
Small proline-rich protein 2A Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
100042514 | |
Research Use Only | |
Sprr2a | |
Recombinant Protein | |
E. coli | |
MSYYQQQCNQ PCRPPPVCPP PKCPEPCPPQ VWPGPCRPVM CFEPCLPSVW PGPCRPVVCY EQCPPQPWQS TCPPVQFPPC QQK | |
Greater than 90% as determined by SDS-PAGE. |