missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TGFB3 Protein
A cDNA sequence encoding the TGFB3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1450.00 €
Specifications
Gene ID (Entrez) | 21809 |
---|---|
Name | TGFB3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Tgfb3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975438
|
enQuireBio™
QP10876-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965438
|
enQuireBio™
QP10876-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955438
|
enQuireBio™
QP10876-100UG |
100 μg |
1450.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
21809 | |
Research Use Only | |
Tgfb3 | |
Recombinant Protein | |
E. coli | |
ALDTNYCFRN LEENCCVRPL YIDFRQDLGW KWVHEPKGYY ANFCSGPCPY LRSADTTHST VLGLYNTLNP EASASPCCVP QDLEPLTILY YVGRTPKVEQ LSNMVVKSCK CS | |
Greater than 96.0% as determined by SDS-PAGE. |
TGFB3 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity is determined by the ability to induce chondrogenic differentiation. | |
Mouse | |
Untagged | |
The Mouse TGFB3 protein solution contains 20% Ethanol and 0.12% Acetic acid. |