missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TGFB3 Protein
Recombinant Protein
4325.00 € - 7105.00 €
Specifications
Accession Number | NP_001316867.1 |
---|---|
Gene ID (Entrez) | 7043 |
Name | TGFB3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986109
|
enQuireBio™
QP5430-500UG |
500 μg |
4325.00 €
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976109
|
enQuireBio™
QP5430-1MG |
1 mg |
7105.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001316867.1 | |
TGFB3 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS | |
Greater than 95% as determined by SDS-PAGE. |
7043 | |
Research Use Only | |
TGFB3 | |
Mouse | |
Untagged | |
10 mM acetic acid and 20% Ethanol at a concentration of 0.25 mg/mL |