missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TLR2 Protein
A cDNA sequence encoding the TLR2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | TLR2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974609
|
enQuireBio™
QP13759-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994609
|
enQuireBio™
QP13759-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984609
|
enQuireBio™
QP13759-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TLR2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
TLR2 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLQESLSCD ASGVCDGRSR SFTSIPSGLT AAMKSLDLSF NKITYIGHGD LRACANLQVL MLKSSRINTI EGDAFYSLGS LEHLDLSDNH LSSLSSSWFG PLSSLKYLNL MGNPYQTLGV TSLFPNLTNL QTLRIGNVET FSEIRRIDFA GLTSLNELEI KALSLRNYQS QSLKSIRDIH HLTLHLSESA FLLEIFADIL SSVRYLELRD TNLARFQFSP LPVDEVSSPM KKLAFRGSVL TDESFNELLK LLRYILELSE VEFDDCTLNG LGDFNPSESD VVSELGKVET VTIRRLHIPQ FYLFYDLSTV YSLLEKVKRI TVENSKVFLV PCSFSQHLKS LEFLDLSENL MVEEYLKNSA CKGAWPSLQT LVLSQNHLRS MQKTGEILLT LKNLTSLDIS RNTFHPMPDS CQWPEKMRFL NLSSTGIRVV KTCIPQTLEV LDVSNNNLDS FSLFLPRLQE LYISRNKLKT LPDASLFPVL LVMKIRENAV STFSKDQLGS FPKLETLEAG DNHFVCSCEL LSFTMETPAL AQILVDWPDS YLCDSPPRLH GHRLQDARPS VLECHQLEPK SCDKTHTCPP CPAPELLGGP SVFLFPPKPK DTLMISRTPE VTCVVVDVSH EDPEVKFNWY VDGVEVHNAK TKPREEQYNS TYRVVSVLTV LHQDWLNGKE YKCKVSNKAL PAPIEKTISK AKGQPREPQV YTLPPSRDEL TKNQVSLTCL VKGFYPSDIA VEWESNGQPE NNYKTTPPVL DSDGSFFLYS KLTVDKSRWQ QGNVFSCSVM HEALHNHYTQ KSLSLSPGKH HHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |